Poly(acrylic acid), 25 wt% soln. in water
Poly(acrylic acid), 25 wt% soln. in water
Poly(acrylic acid), 25 wt% soln. in water
Thermo Scientific Chemicals

Poly(acrylic acid), 25 wt% soln. in water

CAS: 9003-01-4 | (C3H4O2)n | 72.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number 044669.36
also known as 044669-36
Price (EUR)
216,00
Each
Quantity:
500 g
Request bulk or custom format
Price (EUR)
216,00
Each
Chemical Identifiers
CAS9003-01-4
IUPAC NamePoly(acrylic acid)
Molecular Formula(C3H4O2)n
SMILESOC(=O)C(-*)C-*
MDL NumberMFCD00084394
SpecificationsSpecification SheetSpecification Sheet
Assay from Suppliers CofA23.0-27.0% w/w [Poly(acrylic acid)]
ClarityClear
Identification (FTIR)Conforms
FormLiquid
Polyacrylic acid is used in disposable diapers and in ion exchange resins. It is also used to study solute diffusion in polyvinyl alcohol/polyacrylic acid copolymer hydrogel. It is also employed as a thickening, suspending, emulsifying and dispersing agent in pharmaceuticals, cosmetics, adhesives and paints. Further, it is used for the preparation of poly(N-isopropylacrylamide)-block-polyacrylic acid copolymer which responds to both temperature and pH stimuli. In addition to this, it is used in preparing block copolymer of oligo (methyl methacrylate)/PAA for micellar delivery of hydrophobic drugs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Frequently asked questions (FAQs)

What is the average molecular weight of Poly(acrylic acid), 25 wt% soln. in water?

The average molecular weight of the Poly(acrylic acid), 25 wt% soln. in water is 240,000 g/mole.