Palladium nitrate, Matrix Modifier Solution, Specpure™
Thermo Scientific Chemicals

Palladium nitrate, Matrix Modifier Solution, Specpure™

 
Have Questions?
Catalog NumberQuantity
044765.AE
also known as 044765-AE
100 mL
Catalog number 044765.AE
also known as 044765-AE
Price (EUR)
228,65
온라인 행사
254,00
Save 25,35 (10%)
Each
Quantity:
100 mL
Request bulk or custom format
Price (EUR)
228,65
온라인 행사
254,00
Save 25,35 (10%)
Each
Specifications
Chemical Name or MaterialPalladium nitrate Matrix Modifier Solution
DOT InformationTransport Hazard Class: 8; Packing Group: II; Proper Shipping Name: NITRIC ACID
EINECS Number231-714-2
Health Hazard 1H290-H314-H335
Health Hazard 2GHS H Statement
H314
Causes severe skin burns and eye damage.
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only