12-Hydroxystearic acid, 95%
12-Hydroxystearic acid, 95%
12-Hydroxystearic acid, 95%
Thermo Scientific Chemicals

12-Hydroxystearic acid, 95%

CAS: 106-14-9 | C18H36O3 | 300.483 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number 044810.03
also known as 044810-03
Price (EUR)
35,10
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
35,10
Each
Specifications
Beilstein1726730
CAS106-14-9
ChEBICHEBI:85208
Chemical Name or Material12-Hydroxystearic acid
ColorWhite
View more
12-Hydroxystearic acid is used in cosmetics, wax blends, heavy duty greases, polishes, inks and hot melt adhesives, as a lubricant for natural and synthetic rubbers and as a source for industrial oleo chemicals. It is also used as an intermediate in pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only