Iron rod, 7.62mm (0.30in) dia, 99.95% (metals basis)
Iron rod, 7.62mm (0.30in) dia, 99.95% (metals basis)
Iron rod, 7.62mm (0.30in) dia, 99.95% (metals basis)
Thermo Scientific Chemicals

Iron rod, 7.62mm (0.30in) dia, 99.95% (metals basis)

CAS: 7439-89-6 | Fe | 55.85 g/mol
Have Questions?
Catalog NumberQuantity
044918.DQ
also known as 044918-DQ
190 mm
Catalog number 044918.DQ
also known as 044918-DQ
Price (EUR)
161,00
Each
Quantity:
190 mm
Request bulk or custom format
Price (EUR)
161,00
Each
Specifications
AppearanceGray rod
Assay Percent Range99.95% (Metals basis)
CAS7439-89-6
ChEBICHEBI:82664
Chemical Name or MaterialIron
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only