Silver conductive coating
Thermo Scientific Chemicals

Silver conductive coating

 
Have Questions?
Catalog NumberQuantity
044937.1015 g
Catalog number 044937.10
Price (EUR)
-
Quantity:
15 g
Request bulk or custom format
Specifications
Chemical Name or MaterialSilver conductive coating
DOT InformationTransport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ADHESIVES
Health Hazard 1H225-H301+H311-H330
Health Hazard 2GHS H Statement
H225-H300-H310-H330
Highly flammable liquid and vapor.
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Health Hazard 3P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P284-P301+P310-P303+P361+P353-P304+P340-P310-P312-P330-P363-P370+P378q-P501c
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only