Chlorobis(cyclooctene)iridium(I) Dimer, Ir nominally 42.9%
Chlorobis(cyclooctene)iridium(I) Dimer, Ir nominally 42.9%
Chlorobis(cyclooctene)iridium(I) Dimer, Ir nominally 42.9%
Thermo Scientific Chemicals

Chlorobis(cyclooctene)iridium(I) Dimer, Ir nominally 42.9%

CAS: 12246-51-4 | C32H56Cl2Ir2-2 | 896.134 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 044966.03
also known as 044966-03
Price (EUR)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS12246-51-4
IUPAC Nametetrakis(cyclooctene) diiridium dichloride
Molecular FormulaC32H56Cl2Ir2
InChI KeyCJJIQMGSHWWMCK-UHFFFAOYSA-L
SMILES[Cl-].[Cl-].[Ir].[Ir].C1CCCC=CCC1.C1CCCC=CCC1.C1CCCC=CCC1.C1CCCC=CCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)41.90-43.90% Carbon
Assay (unspecified)5.80-6.80% Hydrogen
Assay (unspecified)6.90-8.90% Chlorine
Chlorobis(cyclooctene)iridium(I) dime is used as catalyst for Isomerization-hydroboration reactions with nido-carboranyldiphosphine as stabilizing ligand, Hydrogen peroxide oxidation of hydroxamic acids and their subsequent hetero Diels-Alder cycloaddition reactions, Immobilization of organic functional groups onto solid supports through vinylsilane coupling reactions, Alkylation reactions, Guerbet reaction, Allylic amination reactions in a DNA-diene-iridium(I) hybrid system, Asymmetric hydroamination reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only