Gold wire, 0.35mm (0.0138in) dia, Premion™, 99.999% (metals basis)
Gold wire, 0.35mm (0.0138in) dia, Premion™, 99.999% (metals basis)
Gold wire, 0.35mm (0.0138in) dia, Premion™, 99.999% (metals basis)
Thermo Scientific Chemicals

Gold wire, 0.35mm (0.0138in) dia, Premion™, 99.999% (metals basis)

Form: Approx.1.82g/m | CAS: 7440-57-5 | Au | 196.97 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 cm
100 cm
500 cm
Catalog number 045084.CC
also known as 045084-CC
Price (EUR)
2.448,00
Each
Quantity:
500 cm
Request bulk or custom format
Price (EUR)
2.448,00
Each
Specifications
AppearanceGold colored wire
Assay Percent Range99.999% (Metals basis)
CAS7440-57-5
ChEBICHEBI:30050
Chemical Name or MaterialGold
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only