Bismuth, plasma standard solution, Specpure™ Bi 10μg/mL
Bismuth, plasma standard solution, Specpure™ Bi 10μg/mL
Bismuth, plasma standard solution, Specpure™ Bi 10μg/mL
Thermo Scientific Chemicals

Bismuth, plasma standard solution, Specpure™ Bi 10μg/mL

 
Have Questions?
Catalog NumberQuantity
045249.AE
also known as 045249-AE
100 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 045249.AE
also known as 045249-AE
Price (EUR)
-
Quantity:
100 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialBismuth, plasma standard solution
DOT InformationTransport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S.
EINECS Number231-714-2
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only