Palladium nitrate & Magnesium nitrate, Matrix Modifier Solution, Specpure™
Thermo Scientific Chemicals

Palladium nitrate & Magnesium nitrate, Matrix Modifier Solution, Specpure™

0.1% Pd and 0.06% Mg(NO3)2 in 2% HNO3
Have Questions?
Catalog NumberQuantityPackaging
045291.AK
also known as 045291-AK
250 mLPlastic bottle
Catalog number 045291.AK
also known as 045291-AK
Price (EUR)
189,00
Each
Quantity:
250 mL
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
189,00
Each
Specifications
Chemical Name or MaterialPalladium nitrate & Magnesium nitrate
DOT InformationHazard Class: 8; Packaging Group: III
Health Hazard 1H290-H315-H319
Health Hazard 3P234-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P390
MDL NumberMFCD09910543
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only