D-Glucose dehydrogenase, Thermo Scientific Chemicals
Thermo Scientific Chemicals

D-Glucose dehydrogenase, Thermo Scientific Chemicals

 
Have Questions?
Catalog NumberQuantity
045402.OAT
also known as 045402-O@
5 kU
Catalog number 045402.OAT
also known as 045402-O@
Price (EUR)
667,00
Each
Quantity:
5 kU
Request bulk or custom format
Price (EUR)
667,00
Each
Chemical Identifiers
CAS9028-53-9
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormPowder
Activity (U)7.2
Method1 unit will reduce 1.0µmol of NAD to NADH per minute at pH 7, 25°C while osidising glucose (100mM) to glucon0-1,5-lactone
Diagnostics and cofactor regeneration of NAD(P)H in reductive processes

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only