Carbon, activated, -20+40 mesh
Carbon, activated, -20+40 mesh
Carbon, activated, -20+40 mesh
Thermo Scientific Chemicals

Carbon, activated, -20+40 mesh

CAS: 7440-44-0 | C | 12.011 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2 kg
Catalog number 045478.A3
also known as 045478-A3
Price (EUR)
150,00
Each
Quantity:
2 kg
Request bulk or custom format
Price (EUR)
150,00
Each
Chemical Identifiers
CAS7440-44-0
IUPAC Namecarbon
Molecular FormulaC
InChI KeyOKTJSMMVPCPJKN-UHFFFAOYSA-N
SMILES[C]
View more
SpecificationsSpecification SheetSpecification Sheet
Particle Size-20+40 mesh (425-850 micron)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only