Gold nanopowder, APS 20-25nm, 99.95+% (metals basis)
Gold nanopowder, APS 20-25nm, 99.95+% (metals basis)
Gold nanopowder, APS 20-25nm, 99.95+% (metals basis)
Thermo Scientific Chemicals

Gold nanopowder, APS 20-25nm, 99.95+% (metals basis)

CAS: 7440-57-5 | Au | 196.97 g/mol
Have Questions?
Catalog NumberQuantity
045822.03
also known as 045822-03
1 g
Catalog number 045822.03
also known as 045822-03
Price (EUR)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS7440-57-5
IUPAC Namegold
Molecular FormulaAu
InChI KeyPCHJSUWPFVWCPO-UHFFFAOYSA-N
SMILES[Au]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.05% max.
Gold nanopowder is intercalated within a polymer matrix lower the effective conjugation length of the polymer, resulting in a blue shift in the adsorption. It also finds application in plasmonics, electronics and optical applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only