Neodymium powder, -200 mesh, 99.9% (REO)
Neodymium powder, -200 mesh, 99.9% (REO)
Neodymium powder, -200 mesh, 99.9% (REO)
Thermo Scientific Chemicals

Neodymium powder, -200 mesh, 99.9% (REO)

CAS: 7440-00-8 | Nd | 144.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 045914.06
also known as 045914-06
Price (EUR)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS7440-00-8
IUPAC Nameneodymium
Molecular FormulaNd
InChI KeyQEFYFXOXNSNQGX-UHFFFAOYSA-N
SMILES[Nd]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Packaged under Argon
RUO – Research Use Only