Copper(II) ethoxide, 98%
Copper(II) ethoxide, 98%
Copper(II) ethoxide, 98%
Thermo Scientific Chemicals

Copper(II) ethoxide, 98%

CAS: 2850-65-9 | C4H10CuO2 | 153.668 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 046316.14
also known as 046316-14
Price (EUR)
513,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
513,00
Each
Chemical Identifiers
CAS2850-65-9
IUPAC Namecopper(2+) diethanolate
Molecular FormulaC4H10CuO2
InChI KeyCRCKGIUJMFFISH-UHFFFAOYSA-N
SMILES[Cu++].CC[O-].CC[O-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Blue to blue-green to green
Assay from Supplier's CofA≥97.5%
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Packaged under Argon
RUO – Research Use Only