Non-wetting Pt 5% Au crucible with RF rim, Top Dia 32.5mm,  Bot Dia 18mm, Ht 36mm, Base Thickness 0.25mm, Capacity 20mL, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Non-wetting Pt 5% Au crucible with RF rim, Top Dia 32.5mm, Bot Dia 18mm, Ht 36mm, Base Thickness 0.25mm, Capacity 20mL, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
046558.KT
also known as 046558-KT
1 Ea.
Catalog number 046558.KT
also known as 046558-KT
Price (EUR)
-
Quantity:
1 Ea.
Request bulk or custom format
Specifications
Quantity1 Ea.