Platinum Cathode mesh cylinder, cyl. Dia 40mm, Ht 40mm, overall Ht 140mm, =S.A. 100cm<sup>2</sup>, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Platinum Cathode mesh cylinder, cyl. Dia 40mm, Ht 40mm, overall Ht 140mm, =S.A. 100cm2, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
046820.KT
also known as 046820-KT
1 Ea.
Catalog number 046820.KT
also known as 046820-KT
Price (EUR)
-
Quantity:
1 Ea.
Request bulk or custom format
Specifications
Quantity1 Ea.