Lanthanum(III) iodide, ultra dry, 99.99% (metals basis)
Lanthanum(III) iodide, ultra dry, 99.99% (metals basis)
Lanthanum(III) iodide, ultra dry, 99.99% (metals basis)
Thermo Scientific Chemicals

Lanthanum(III) iodide, ultra dry, 99.99% (metals basis)

CAS: 13813-22-4 | I3La | 519.62 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 047184.06
also known as 047184-06
Price (EUR)
255,00
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
255,00
Each
Chemical Identifiers
CAS13813-22-4
IUPAC Namelanthanum(3+) triiodide
Molecular FormulaI3La
InChI KeyKYKBXWMMXCGRBA-UHFFFAOYSA-K
SMILES[I-].[I-].[I-].[La+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to Gray
FormPowder
Total Metal Impurities0.01% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Ampouled under argon
RUO – Research Use Only