Bismuth(III) selenide, 99.995% (metals basis)
Bismuth(III) selenide, 99.995% (metals basis)
Bismuth(III) selenide, 99.995% (metals basis)
Thermo Scientific Chemicals

Bismuth(III) selenide, 99.995% (metals basis)

CAS: 12068-69-8
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 047198.03
also known as 047198-03
Price (EUR)
49,70
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
49,70
Each
Chemical Identifiers
CAS12068-69-8
IUPAC Namedibismuth(3+) triselandiide
Molecular FormulaBi2Se3
InChI KeyFBGGJHZVZAAUKJ-UHFFFAOYSA-N
SMILES[Se--].[Se--].[Se--].[Bi+3].[Bi+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)grey to Black to silver
Formgranules
Total Metal Impurities0.005% max.
It is used as a semiconductor thermoelectric material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only