Silver chloride, 4.9% in H{2}O, nanoparticle dispersion, <200nm APS Number Weighted
Silver chloride, 4.9% in H{2}O, nanoparticle dispersion, <200nm APS Number Weighted
Silver chloride, 4.9% in H{2}O, nanoparticle dispersion, <200nm APS Number Weighted
Thermo Scientific Chemicals

Silver chloride, 4.9% in H{2}O, nanoparticle dispersion, <200nm APS Number Weighted

CAS: 7783-90-6 | AgCl | 143.32 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number 047280.22
also known as 047280-22
Price (EUR)
-
Quantity:
100 g
Request bulk or custom format
Specifications
CAS7783-90-6
Chemical Name or MaterialSilver chloride
ColorWhite
Concentration or Composition (by Analyte or Components)4.9% in H2O
DOT InformationTransport Hazard Class: 9; Packaging Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
View more
Used as pharmaceutical intermediate and in pharmaceutical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only