1,4-Dioxane, anhydrous, 99.8%, stab. with 1-3ppm BHT, packaged under argon in resealable AcroSeal™ bottles
1,4-Dioxane, anhydrous, 99.8%, stab. with 1-3ppm BHT, packaged under argon in resealable AcroSeal™ bottles
1,4-Dioxane, anhydrous, 99.8%, stab. with 1-3ppm BHT, packaged under argon in resealable AcroSeal™ bottles
Thermo Scientific Chemicals

1,4-Dioxane, anhydrous, 99.8%, stab. with 1-3ppm BHT, packaged under argon in resealable AcroSeal™ bottles

CAS: 123-91-1 | C{4}H{8}O{2}
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
4 x 1 L
Catalog number 047305.K2
also known as 047305-K2
Price (EUR)
118,00
Each
Quantity:
1 L
Price (EUR)
118,00
Each
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Boiling Point100°C to 102°C
CAS123-91-1
Melting Point11.8°C
Molecular FormulaC{4}H{8}O{2}
Percent Purity99.8%
Quantity1 L
Unit SizeEach