Lithium chromate, anhydrous, 97+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Lithium chromate, anhydrous, 97+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
2.5 kg
Catalog number 047326.30
also known as 047326-30
Price (EUR)
110,00
Each
Quantity:
250 g
Request bulk or custom format
Price (EUR)
110,00
Each
Specifications
CAS14307-35-8
Molecular FormulaCrLi{2}O{4}
Percent Purity≥97%
Physical FormSolid
Quantity250 g
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.