Samarium(II) iodide, ultra dry, 99.99% (REO)
Thermo Scientific Chemicals

Samarium(II) iodide, ultra dry, 99.99% (REO)

CAS: 32248-43-4 | SmI{2}
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 047328.03
Price (EUR)
87,00
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
87,00
Each
Specifications
CAS32248-43-4
Molecular FormulaSmI{2}
Percent Purity99.99%
Physical FormSolid
Quantity1 g
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.