Platinum black, high surface area
Platinum black, high surface area
Platinum black, high surface area
Thermo Scientific Chemicals

Platinum black, high surface area

CAS: 7440-06-4
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 047349.03
also known as 047349-03
Price (EUR)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS7440-06-4
IUPAC Nameplatinum
Molecular FormulaPt
InChI KeyBASFCYQUMIYNBI-UHFFFAOYSA-N
SMILES[Pt]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Black
FormPowder
Assay (unspecified)> 99% Pt
Surface Area>25.0m²/g ( Electrochemical SA)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.