Tetrahydrofuran, anhydrous, 99.8+%, stab. with 0.025% BHT, packaged under Argon in resealable AcroSeal™ bottles
Thermo Scientific™

Tetrahydrofuran, anhydrous, 99.8+%, stab. with 0.025% BHT, packaged under Argon in resealable AcroSeal™ bottles

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
4 x 1 L
Catalog number 047382.K2
also known as 047382-K2
Price (EUR)
85,65
Online Exclusive
94,50
Save 8,85 (9%)
Each
Quantity:
1 L
Request bulk or custom format
Price (EUR)
85,65
Online Exclusive
94,50
Save 8,85 (9%)
Each
Chemical Identifiers
CAS109-99-9
IUPAC Nameoxolane
Molecular FormulaC4H8O
InChI KeyWYURNTSHIVDZCO-UHFFFAOYSA-N
SMILESC1CCOC1
View more
SpecificationsSpecification SheetSpecification Sheet
StabilizerBHT (report only)
Appearance (Color)Colorless
Assay (GC)≥99.8% (corrected for water)
Water Content (Karl Fischer Titration)≤50ppm
FormLiquid