Lithium dichromate hydrate, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Lithium dichromate hydrate, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
2.5 kg
Catalog number 047394.A4
also known as 047394-A4
Price (EUR)
321,00
Each
Quantity:
2.5 kg
Request bulk or custom format
Price (EUR)
321,00
Each
Specifications
CAS13843-81-7
Molecular FormulaCr{2}Li{2}O{7}^.xH{2}O
SynonymChromic acid dilithium salt;
Percent Purity≥98%
Physical FormSolid
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.