Nickel(II) bromide, anhydrous, 99%
Nickel(II) bromide, anhydrous, 99%
Nickel(II) bromide, anhydrous, 99%
Thermo Scientific Chemicals

Nickel(II) bromide, anhydrous, 99%

CAS: 13462-88-9 | Br2Ni | 218.501 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number 053115.18
also known as 053115-18
Price (EUR)
119,00
Each
Quantity:
50 g
Request bulk or custom format
Price (EUR)
119,00
Each
Chemical Identifiers
CAS13462-88-9
IUPAC Namenickel(2+) dibromide
Molecular FormulaBr2Ni
InChI KeyIPLJNQFXJUCRNH-UHFFFAOYSA-L
SMILES[Ni++].[Br-].[Br-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow-brown to orange-brown to brown
FormPowder or chunks
Assay (unspecified)≥98.5%
Nickel(II) bromide is used as a source of the bromide ion. It is an important raw material and intermediate used in synthetic chemistry. Further, it is used as a catalyst in cross-coupling reactions.It is used in the preparation of paramagnetic trans-diaquabis(N,N-diethylethylenediamine) nickel(II) bromide and diamagnetic bis(N,N-diethylethylenediamine)nickel(II) bromide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only