Scandium(III) nitrate hydrate, REacton™, 99.9% (REO)
Scandium(III) nitrate hydrate, REacton™, 99.9% (REO)
Scandium(III) nitrate hydrate, REacton™, 99.9% (REO)
Scandium(III) nitrate hydrate, REacton™, 99.9% (REO)
Thermo Scientific Chemicals

Scandium(III) nitrate hydrate, REacton™, 99.9% (REO)

CAS: 107552-14-7 | N3O9Sc | 230.97 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 067104.06
Price (EUR)
463,00
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (EUR)
463,00
Each
Add to cart
Chemical Identifiers
CAS107552-14-7
IUPAC Namescandium(3+) trinitrate
Molecular FormulaN3O9Sc
InChI KeyDFCYEXJMCFQPPA-UHFFFAOYSA-N
SMILES[Sc+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.
FormPowder or crystal or chunk(s)
Appearance (Color)White to pale cream
Scandium(III) nitrate hydrate is used as an oxidizing agent in organic synthesis. It is used in the optical coatings, electronic ceramics and laser industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only