Titanium(IV) ethoxide, TiO{2} 33% min
Titanium(IV) ethoxide, TiO{2} 33% min
Titanium(IV) ethoxide, TiO{2} 33% min
Thermo Scientific Chemicals

Titanium(IV) ethoxide, TiO{2} 33% min

CAS: 3087-36-3 | C8H20O4Ti | 228.111 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
50 g
250 g
Catalog number 077142.06
also known as 077142-06
Price (EUR)
35,90
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
35,90
Each
Chemical Identifiers
CAS3087-36-3
IUPAC Nametitanium(4+) tetraethanolate
Molecular FormulaC8H20O4Ti
InChI KeyJMXKSZRRTHPKDL-UHFFFAOYSA-N
SMILES[Ti+4].CC[O-].CC[O-].CC[O-].CC[O-]
View more
SpecificationsSpecification SheetSpecification Sheet
Formviscous liquid/ fused solidified melt
Assay (unspecified)TiO₂ 33%, min
Appearance (Color)Colorless to yellow
Titanium(IV) ethoxide is used in organic synthesis and material science. It is involved in the synthesis of anti-perspirants and antifungal agents. Furthermore, it is used to prepare carbon films in association with silicon dioxide. It is also used as a precursor for the synthesis of Ti4+ ions titanium nanoparticles by various methods.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only