Cyclopentadienylvanadium tetracarbonyl, 97+%
Cyclopentadienylvanadium tetracarbonyl, 97+%
Cyclopentadienylvanadium tetracarbonyl, 97+%
Thermo Scientific Chemicals

Cyclopentadienylvanadium tetracarbonyl, 97+%

CAS: 12108-04-2 | C9H5O4V | 228.076 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
0.5 g
2 g
Catalog number 081123.02
also known as 081123-02
Price (EUR)
112,00
Each
Quantity:
0.5 g
Request bulk or custom format
Price (EUR)
112,00
Each
Specifications
CAS12108-04-2
Chemical Name or MaterialCyclopentadienylvanadium tetracarbonyl
DOT InformationTransport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: METAL CARBONYLS, SOLID, N.O.S.
EINECS Number235-163-9
Formula Weight228.08
View more
Employed as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only