This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 088053.AE
also known as 088053-AE
Price (EUR)
-
Quantity:
100 mL
Request bulk or custom format
Specifications
CAS19049-40-2
CAS7732-18-5
Chemical Name or MaterialBeryllium
ColorColorless
Concentration or Composition (by Analyte or Components)Nitric acid: 5%; Beryllium oxide acetate: 1.13%; Water: 93.87%
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.