Magnesium boride, 99% (metals basis)
Magnesium boride, 99% (metals basis)
Magnesium boride, 99% (metals basis)
Thermo Scientific Chemicals

Magnesium boride, 99% (metals basis)

CAS: 12007-25-9 | MgB2
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number 088149.14
also known as 088149-14
Price (EUR)
159,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
159,00
Each
Chemical Identifiers
CAS12007-25-9
IUPAC Namediboron magnesium
Molecular FormulaB2Mg
InChI KeyPZKRHHZKOQZHIO-UHFFFAOYSA-N
SMILES[B].[B].[Mg]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities1% max.
Magnesium Boride exhibits super conducting properties and utilized in MRI superconducting magnet, which is a closed-loop cryocooler. It is used as propellants, pyrotechnics and explosives. It can be used as high temperature boron-based thermoelectric materials. It is also used in superconducting transformers, rotors, transmission cables, superconducting radio frequency cavities to minimize energy loss.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only