Copper Phosphorus shot
Thermo Scientific Chemicals

Copper Phosphorus shot

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2 kg
Catalog number 088357.36
also known as 088357-36
Price (EUR)
-
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialCopper Phosphorus shot
MDL NumberMFCD01074956
Recommended StorageAmbient temperatures
TSCAYes
Quantity500 g
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only