Titanium(diisopropoxide) bis(2,4-pentanedionate), 75% in isopropanol
Titanium(diisopropoxide) bis(2,4-pentanedionate), 75% in isopropanol
Titanium(diisopropoxide) bis(2,4-pentanedionate), 75% in isopropanol
Thermo Scientific Chemicals

Titanium(diisopropoxide) bis(2,4-pentanedionate), 75% in isopropanol

CAS: 17927-72-9 | C16H28O6Ti
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number 089360.22
also known as 089360-22
Price (EUR)
41,00
Each
Quantity:
100 g
Request bulk or custom format
Price (EUR)
41,00
Each
Chemical Identifiers
CAS17927-72-9
IUPAC Nametitanium(4+) bis(2,4-dioxopentan-3-ide) bis(propan-2-olate)
Molecular FormulaC16H28O6Ti
InChI KeyDOOBMVWXHWRGEB-UHFFFAOYSA-N
SMILES[Ti+4].CC(C)[O-].CC(C)[O-].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear pale brown to brown
FormLiquid
Assay (unspecified)≥9.50% Ti
Titanium(diisopropoxide) bis(2,4-pentanedionate) is used as catalyst, used in sol-gel, coating, primer and crosslinking.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only