S-Acetylthiocholine iodide, 98%
S-Acetylthiocholine iodide, 98%
S-Acetylthiocholine iodide, 98%
Thermo Scientific Chemicals

S-Acetylthiocholine iodide, 98%

CAS: 1866-15-5 | C7H16INOS | 289.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 102710050
Price (EUR)
129,00
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
129,00
Each
Chemical Identifiers
CAS1866-15-5
IUPAC Name[2-(acetylsulfanyl)ethyl]trimethylazanium iodide
Molecular FormulaC7H16INOS
InChI KeyNTBLZMAMTZXLBP-UHFFFAOYSA-M
SMILES[I-].CC(=O)SCC[N+](C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to almost white
Appearance (Form)Powder or crystals
Infrared spectrumConforms
Titration Argentometric97.5 to 102.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only