5-Amino-2-chlorobenzotrifluoride, 99%
5-Amino-2-chlorobenzotrifluoride, 99%
5-Amino-2-chlorobenzotrifluoride, 99%
Thermo Scientific Chemicals

5-Amino-2-chlorobenzotrifluoride, 99%

CAS: 320-51-4 | C7H5ClF3N | 195.57 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 103370050
Price (EUR)
23,50
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
23,50
Each
Chemical Identifiers
CAS320-51-4
IUPAC Name4-chloro-3-(trifluoromethyl)aniline
Molecular FormulaC7H5ClF3N
InChI KeyASPDJZINBYYZRU-UHFFFAOYSA-N
SMILESNC1=CC=C(Cl)C(=C1)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pink to light orange
Infrared spectrumConforms
GC>=98.5 %
Melting point34°C to 38°C
Appearance (Form)Crystalline powder or low melting solid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only