2-Aminopyridine, 99+%
2-Aminopyridine, 99+%
2-Aminopyridine, 99+%
Thermo Scientific Chemicals

2-Aminopyridine, 99+%

CAS: 504-29-0 | C5H6N2 | 94.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Catalog number 104541000
Price (EUR)
38,65
Online exclusive
42,50
Save 3,85 (9%)
Each
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
38,65
Online exclusive
42,50
Save 3,85 (9%)
Each
Chemical Identifiers
CAS504-29-0
IUPAC Namepyridin-2-amine
Molecular FormulaC5H6N2
InChI KeyICSNLGPSRYBMBD-UHFFFAOYSA-N
SMILESNC1=CC=CC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point55°C to 60°C
GC>=99.0 %
Appearance (Color)White to yellow to light beige
Infrared spectrumConforms
Appearance (Form)Powder or crystals or flakes and/or chunks
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only