1,4-Benzoquinone, 98+%
1,4-Benzoquinone, 98+%
1,4-Benzoquinone, 98+%
Thermo Scientific Chemicals

1,4-Benzoquinone, 98+%

1, 4-Benzoquinone, 99%, C6H4O2, CAS Number-106-51-4, 1,4-benzoquine, 1,4-benzoquinone, 2,5-cyclohexadiene-1,4-dione, benzoquinone, chinone, cyclohexadienedione, para-quinone, p-benzoquinone, p-quinone, quinone, 1kg, 98.5% min. (Iodometry), 07,609, 100.0, CHEBI:16509, Yellow to Green, 1.3100g/mL | CAS: 106-51-4 | C6H4O2 | 108.1 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
1 kg
5 kg
25 kg
Packaging:
View all
Plastic drum
Catalog number 105630051
Price (EUR)
714,00
Each
Quantity:
5 kg
Packaging:
Plastic drum
Request bulk or custom format
Price (EUR)
714,00
Each
Chemical Identifiers
CAS106-51-4
IUPAC Namecyclohexa-2,5-diene-1,4-dione
Molecular FormulaC6H4O2
InChI KeyAZQWKYJCGOJGHM-UHFFFAOYSA-N
SMILESO=C1C=CC(=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystals
Infrared spectrumConforms
Melting point110°C to 116°C
Titration Iodometric98.0 to 102.0 %
Appearance (Color)Yellow to green-brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only