4-Bromobenzaldehyde, 99%
4-Bromobenzaldehyde, 99%
4-Bromobenzaldehyde, 99%
Thermo Scientific Chemicals

4-Bromobenzaldehyde, 99%

CAS: 1122-91-4 | C7H5BrO | 185.02 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 106670250
Price (EUR)
69,90
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
69,90
Each
Chemical Identifiers
CAS1122-91-4
IUPAC Name4-bromobenzaldehyde
Molecular FormulaC7H5BrO
InChI KeyZRYZBQLXDKPBDU-UHFFFAOYSA-N
SMILESBrC1=CC=C(C=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Crystalline powder or crystals and/or
Melting point55°C to 59°C
Appearance (Color)White to light beige
Appearance (Form)chunks or fused solid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only