4-Bromofluorobenzene, 99%
4-Bromofluorobenzene, 99%
4-Bromofluorobenzene, 99%
Thermo Scientific Chemicals

4-Bromofluorobenzene, 99%

CAS: 460-00-4 | C6H4BrF | 175.00 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mL
100 mL
500 mL
Packaging:
Glass bottle
Catalog number 107015000
Price (EUR)
312,00
Each
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
312,00
Each
Chemical Identifiers
CAS460-00-4
IUPAC Name1-bromo-4-fluorobenzene
Molecular FormulaC6H4BrF
InChI KeyAITNMTXHTIIIBB-UHFFFAOYSA-N
SMILESFC1=CC=C(Br)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Appearance (Color)Clear colorless
Infrared spectrumConforms
Refractive index1.5260 to 1.5280 (20°C, 589 nm)
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only