6-Bromo-2-naphthol, 97%
6-Bromo-2-naphthol, 97%
6-Bromo-2-naphthol, 97%
6-Bromo-2-naphthol, 97%
Thermo Scientific Chemicals

6-Bromo-2-naphthol, 97%

CAS: 15231-91-1 | C10H7BrO | 223.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 107171000
Price (EUR)
124,10
Online Exclusive
146,00
Save 21,90 (15%)
Each
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
124,10
Online Exclusive
146,00
Save 21,90 (15%)
Each
Add to cart
Chemical Identifiers
CAS15231-91-1
IUPAC Name6-bromonaphthalen-2-ol
Molecular FormulaC10H7BrO
InChI KeyYLDFTMJPQJXGSS-UHFFFAOYSA-N
SMILESOC1=CC=C2C=C(Br)C=CC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white to beige
Infrared spectrumConforms
Melting point125°C to 130°C
GC>=96.0 %
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only