n-Butylamine, 99+%
n-Butylamine, 99+%
n-Butylamine, 99+%
Thermo Scientific Chemicals

n-Butylamine, 99+%

CAS: 109-73-9 | C4H11N | 73.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
2.5 L
10 L
Packaging:
View all
Metal drum
Catalog number 107800100
Price (EUR)
381,00
Each
Quantity:
10 L
Packaging:
Metal drum
Request bulk or custom format
Price (EUR)
381,00
Each
Chemical Identifiers
CAS109-73-9
IUPAC Namebutan-1-amine
Molecular FormulaC4H11N
InChI KeyHQABUPZFAYXKJW-UHFFFAOYSA-N
SMILESCCCCN
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<20 APHA
Refractive index1.4000 to 1.4020 (20°C, 589 nm)
Appearance (Form)Clear liquid
Infrared spectrumConforms
GC>=99.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only