S-Butyrylthiocholine iodide, 98%
S-Butyrylthiocholine iodide, 98%
S-Butyrylthiocholine iodide, 98%
Thermo Scientific Chemicals

S-Butyrylthiocholine iodide, 98%

CAS: 1866-16-6 | C9H20INOS | 317.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 108150250
Price (EUR)
186,00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
186,00
Each
Chemical Identifiers
CAS1866-16-6
IUPAC Name[2-(butanoylsulfanyl)ethyl]trimethylazanium iodide
Molecular FormulaC9H20INOS
InChI KeyWEQAAFZDJROSBF-UHFFFAOYSA-M
SMILES[I-].CCCC(=O)SCC[N+](C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Infrared spectrumConforms
Titration Argentometric>=97.5 %
Appearance (Form)Crystalline powder or crystals
Melting point172°C to 176°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only