epsilon-Caprolactam, 99+%
epsilon-Caprolactam, 99+%
epsilon-Caprolactam, 99+%
epsilon-Caprolactam, 99+%
Thermo Scientific Chemicals

epsilon-Caprolactam, 99+%

CAS: 105-60-2 | C6H11NO | 113.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
250 g
1 kg
2.5 kg
Packaging:
Plastic bottle
Catalog number 108231000
Price (EUR)
19,65
Online Exclusive
21,20
Save 1,55 (7%)
Each
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
19,65
Online Exclusive
21,20
Save 1,55 (7%)
Each
Add to cart
Chemical Identifiers
CAS105-60-2
IUPAC Nameazepan-2-one
Molecular FormulaC6H11NO
InChI KeyJBKVHLHDHHXQEQ-UHFFFAOYSA-N
SMILESO=C1CCCCCN1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystals or flakes
Melting point68°C to 71°C
Appearance (Color)White
GC>=99.0 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only