Carbamylcholine chloride, 98+%
Carbamylcholine chloride, 98+%
Carbamylcholine chloride, 98+%
Carbamylcholine chloride, 98+%
Thermo Scientific Chemicals

Carbamylcholine chloride, 98+%

CAS: 51-83-2 | C6H15ClN2O2 | 182.65 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 108240250
Price (EUR)
369,00
Each
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
369,00
Each
Add to cart
Chemical Identifiers
CAS51-83-2
IUPAC Name2-(trimethylazaniumyl)ethyl carbamate chloride
Molecular FormulaC6H15ClN2O2
InChI KeyAIXAANGOTKPUOY-UHFFFAOYSA-N
SMILES[Cl-].C[N+](C)(C)CCOC(N)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light cream
Appearance (Form)Crystalline powder or crystals
Infrared spectrumConforms
Melting point200°C to 208°C (Dec)
Titration with HClO498.0 to 102.0 % (On dried substance)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only