2-chloroaniline, 98+%
2-chloroaniline, 98+%
2-chloroaniline, 98+%
Thermo Scientific Chemicals

2-chloroaniline, 98+%

CAS: 95-51-2 | C6H6ClN | 127.57 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
5 L
Packaging:
View all
Glass bottle
Catalog number 108570010
Price (EUR)
83,40
Each
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
83,40
Each
Chemical Identifiers
CAS95-51-2
IUPAC Name2-chloroaniline
Molecular FormulaC6H6ClN
InChI KeyAKCRQHGQIJBRMN-UHFFFAOYSA-N
SMILESNC1=CC=CC=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=98.0 %
AppearanceMay darken on storage
Appearance (Color)Clear light yellow to brown
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only