2-Chlorocyclopentanone, 97%, stabilized
2-Chlorocyclopentanone, 97%, stabilized
2-Chlorocyclopentanone, 97%, stabilized
Thermo Scientific Chemicals

2-Chlorocyclopentanone, 97%, stabilized

CAS: 694-28-0 | C5H7ClO | 118.56 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 108970050
Price (EUR)
52,65
Online exclusive
58,00
Save 5,35 (9%)
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
52,65
Online exclusive
58,00
Save 5,35 (9%)
Each
Chemical Identifiers
CAS694-28-0
SpecificationsSpecification SheetSpecification Sheet
GC>=96.0 %
Refractive index1.4730 to 1.4770 (20°C, 589 nm)
Stabilizer1 % Potassium carbonate
Appearance (Color)Clear light yellow to brown
Appearance (Form)Liquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only