4-(4-Chlorophenyl)-1,2,3,6-tetrahydropyridine hydrochloride, 97%
4-(4-Chlorophenyl)-1,2,3,6-tetrahydropyridine hydrochloride, 97%
4-(4-Chlorophenyl)-1,2,3,6-tetrahydropyridine hydrochloride, 97%
Thermo Scientific Chemicals

4-(4-Chlorophenyl)-1,2,3,6-tetrahydropyridine hydrochloride, 97%

CAS: 51304-61-1 | C11H12ClN·HCl | 230.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 109930250
Price (EUR)
401,00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
401,00
Each
Chemical Identifiers
CAS51304-61-1
IUPAC Namehydrogen 4-(4-chlorophenyl)-1,2,3,6-tetrahydropyridine chloride
Molecular FormulaC11H13Cl2N
InChI KeyNGDJUNABWNTULX-UHFFFAOYSA-N
SMILES[H+].[Cl-].ClC1=CC=C(C=C1)C1=CCNCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Titration with HClO4>=96.0 %
Appearance (Color)White to light yellow
Melting point199°C to 204°C
Appearance (Form)Crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only