4-Cyanobenzoic acid, 99%
4-Cyanobenzoic acid, 99%
4-Cyanobenzoic acid, 99%
Thermo Scientific Chemicals

4-Cyanobenzoic acid, 99%

CAS: 619-65-8 | C8H5NO2 | 147.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 110730250
Price (EUR)
123,00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
123,00
Each
Chemical Identifiers
CAS619-65-8
IUPAC Name4-cyanobenzoic acid
Molecular FormulaC8H5NO2
InChI KeyADCUEPOHPCPMCE-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=C(C=C1)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Melting point217°C to 222°C
Appearance (Color)White to yellow to light beige
HPLC>=98.5 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only