1,3-Diamino-2-hydroxypropane, 95%
1,3-Diamino-2-hydroxypropane, 95%
1,3-Diamino-2-hydroxypropane, 95%
Thermo Scientific Chemicals

1,3-Diamino-2-hydroxypropane, 95%

CAS: 616-29-5 | C3H10N2O | 90.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 112220050
Price (EUR)
53,50
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
53,50
Each
Chemical Identifiers
CAS616-29-5
IUPAC Name1,3-diaminopropan-2-ol
Molecular FormulaC3H10N2O
InChI KeyUYBWIEGTWASWSR-UHFFFAOYSA-N
SMILESNCC(O)CN
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
Appearance (Form)Low melting solid
Infrared spectrumConforms
Titration with HClO4>=94 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only