2,4'-Dibromoacetophenone, 98%
2,4'-Dibromoacetophenone, 98%
2,4'-Dibromoacetophenone, 98%
Thermo Scientific Chemicals

2,4'-Dibromoacetophenone, 98%

CAS: 99-73-0 | C8H6Br2O | 277.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
Catalog number 112680500
Price (EUR)
105,00
Each
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
105,00
Each
Chemical Identifiers
CAS99-73-0
IUPAC Name2-bromo-1-(4-bromophenyl)ethan-1-one
Molecular FormulaC8H6Br2O
InChI KeyFKJSFKCZZIXQIP-UHFFFAOYSA-N
SMILESBrCC(=O)C1=CC=C(Br)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Light yellow to beige
Appearance (Form)Crystalline solid
Infrared spectrumConforms
Melting point107°C to 111°C
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only